    Sort by:
  • Search Within Results
Pentostatin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Pentostatin is an irreversible inhibitor of adenosine deaminase (Ki = 2.5 pM). Pentostatin is effective in the treatment of many lymphoproliferat...