    Sort by:
  • Search Within Results
PF-06409577 
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   PF-06409577 is a potent and selective allosteric activator of AMPK (EC₅₀ values are 7 and >40000 nM for AMPKα1β1γ1 and AMPKα1β2γ1, respective...