    Sort by:
  • Search Within Results
Brigatinib analog
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Brigantinib alalog is an analog of Brigatinib (AP26113) (Cat.No. 9675), a phosphine oxide-containing ALK inhibitor. It potently inhibits ALK (IC₅₀&nbs...