    Sort by:
  • Search Within Results
MK-4827 (Tosylate)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   MK-4827 is a potent and selective PARP-1 and PARP-2 inhibitor with IC₅₀ values of 3.8 nM and 2.1 nM, respectively. It induces synthetic lethality in p...