    Sort by:
  • Search Within Results
ML-7 hydrochloride
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   ML-7 hydrochloride is a selective MLCK (myosin light chain kinase) inhibitor (Ki = 0.3 μM). Displays reversible, ATP-competitive inhibition ...