    Sort by:
  • Search Within Results
Firocoxib
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Firocoxib is a non-steroidal anti-inflammatory drug (NSAID) that acts as a potent and selective cyclooxygenase (COX)-2 inhibitor with an IC₅₀ of 0.13 ...