    Sort by:
  • Search Within Results
AMG-925
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   AMG-925 is a potent and orally bioavailable dual FLT3/CDK4 inhibitor with IC₅₀ of 1 nM and 3 nM, respectively. It inhibits the pro...