    Sort by:
ABL-001
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   ABL-001 is a potent allosteric inhibitor of BCR-ABL. ABL001 binds to the myristoyl pocket of ABL1 and induces the formation of an inactive kinase conf...