    Sort by:
  • Search Within Results
A-1331852
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   A-1331852 is a potent anti-apoptotic Bcl-xL protein inhibitor and apoptosis-inducing agent useful in the treatment of cancer, immune and autoimmune di...