    Sort by:
  • Search Within Results
ABT-494
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   ABT-494 is a potent and selective Janus kinase JAK-1 inhibitor (IC₅₀ = 43 nM). ABT-494 is 74-fold more selective for JAK-1 over JAK-2 and 58-fold more...