    Sort by:
  • Search Within Results
MLN-8054
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   MLN-8054 is a potent and selective inhibitor of Aurora kinase A (IC₅₀ = 4 nM). Displays 40-fold selectivity for Aurora A over Aurora B.