    Sort by:
  • Search Within Results
Hesperadin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Hesperadin is a potent, ATP-competitive inhibitor of Aurora B kinase (IC₅₀ = 250 nM). Hesperadin inhibits Aurora B kinase by phosphorylation of histon...