    Sort by:
  • Search Within Results
CCT-137690
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   CCT137690 is a highly selective and orally bioavailable inhibitor of Aurora A, Aurora B and Aurora C with IC₅₀ of 15 nM, 25 nM and 19 nM, respectively...