    Sort by:
  • Search Within Results
CCT-129202
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   CCT-129202 is a potent, ATP-competitive inhibitor of Aurora kinases with IC₅₀ values of 42 nM, 198 nM and 227 nM for Aurora A, B and C, respectively. ...