    Sort by:
  • Search Within Results
GDC-0917
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   GDC-0917 (CUDC-427) is a second mitochondrial activator of caspases (Smac) mimetic, a small molecule inhibitor, that mimics Smac, an endogenous antago...