    Sort by:
  • Search Within Results
OSMI-1
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   OSMI-1 is a potent, cell-permeable, O-GlcNAc transferase (OGT) inhibitor.  inhibits protein O-GlcNAcylation in several mammalian cell lines witho...