    Sort by:
BMS-345541
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   BMS-345541 is a highly selective inhibitor of IKK-1 and IKK-2 with IC₅₀ value of 4 μM and 0.3 μM, respectively. It binds at an allosteric site of the ...