    Sort by:
  • Search Within Results
trans-ISRIB
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   trans-ISRIB is an integrated stress response inhibitor. Potently reverses the effects of initiation factor 2α (eIF2α) phosphorylation (IC₅₀ = 5 nM). E...