    Sort by:
  • Search Within Results
SL0101
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   SL0101 is a potent and selective inhibitor of ribosomal S6 kinase (RSK) (IC₅₀ = 89 nM for RSK2). Does not have any effect on upstream kinases suc...