    Sort by:
You Searched For:
Elevenostat
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (76269-740)
Supplier:  BioVision
Description:   Elevenostat (JB3-22), is a small molecule hydroxamic acid derivative that acts as a HDAC11-selective inhibitor. Reporter assays using a luciferase-lin...