    Sort by:
  • Search Within Results
U-73343
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   U-73343 is a structural analog of U-73122 (Cat. No. B1887) that does not inhibit PLC, making it useful as a negative control for PLC inhibition. Cell-...