    Sort by:
  • Search Within Results
AZ-505 ditrifluoroacetate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   AZ-505 is a potent and highly selective inhibitor of SMYD2 (IC₅₀ = 0.12 μM) with potential anticancer activity.  Displays >600-fold selectivi...