    Sort by:
  • Search Within Results
CPI-455
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   CPI-455 is a potent and selective KDM5A inhibitor (IC₅₀ ~10 nM). It inhibits KDM5A, KDM5B and KDM5C to similar extents but shows substantially weaker ...