    Sort by:
  • Search Within Results
BG-45
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   BG-45 is a potent HDAC class I inhibitor with selectivity for HDAC3 (IC₅₀ = 289 nM) over HDAC1, 2. BG45 does not inhibit HDAC6. BG-45 alone or in...