    Sort by:
  • Search Within Results
Atglistatin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Atglistatin is a potent, and selective inhibitor of adipose triglyceride lipase (ATGL) the rate limiting enzyme involved in fatty acid mobilization an...