    Sort by:
TMP-195
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   TMP-195 is a potent and selective class IIa HDAC inhibitor with IC₅₀ values of 59, 60, 26, and 15 nM for HDAC4, HDAC5, HDAC7 and HDAC9 respectively. T...