    Sort by:
  • Search Within Results
Bosentan
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Bosentan is an antagonist of the endothelin receptors type A (Ki = 6.5 nM) and type B (Ki = 340 nM). Inhibits ET-1 induced contraction of is...