    Sort by:
HG-9-91-01
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   HG-9-91-01 is a potent, selective, ATP-competitive, pan-SIK (salt-inducible kinases) inhibitor with IC₅₀ values of 0.92 nM, 6.6n M and 9.6 nM for SIK1...