    Sort by:
  • Search Within Results
UK-371804
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   UK-371804 is a potent urokinase-type plasminogen activator (uPA) inhibitor with a Ki of 10 nM. Displays 4000-fold selectivity vs tPA and 270...