    Sort by:
  • Search Within Results
GJ-103 sodium salt
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   GJ103 sodium salt is an active, water-soluble analog of the read-through compound GJ072. Chemical-induced read through of premature stop codons might ...