    Sort by:
  • Search Within Results
TCS OX2 29
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   TCS OX2 29 is a potent and selective OX2 (Orexin-2) receptor antagonist (IC₅₀ = 40 nM). Displays >250-fold selectivity for OX2 over...