    Sort by:
  • Search Within Results
GLPG0634 Analog
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   GLPG0634 analog is a potent and selective JAK1 inhibitor with IC₅₀ values of 10, 28, 810, and 116 nM for JAK1, JAK2, JAK3, and TYK2, respectively.