    Sort by:
  • Search Within Results
MI-3
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   MI-3 is a Menin-MLL (Mixed linage leukemia) protein interaction inhibitor with an IC₅₀ of 648 ± 25 nM. MLL protein is a histone methyltransferase that...