    Sort by:
  • Search Within Results
A-1155463
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   A-1155463 is a highly potent and selective BCL-XL inhibitor. A-1155463 is more potent against BCL-XL-dependent cell lines relative to WEHI-539 (Cat. N...