    Sort by:
  • Search Within Results
BVT948
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   BVT 948 is a noncompetitive, cell-permeable inhibitor of protein tyrosine phosphatases (PTP; IC₅₀s = 0.09-1.7 µM). displays irreversible inhibition th...