You Searched For: 4-Methylumbelliferyl-alpha-D-glucopyranoside
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (80058-382)
Supplier: MilliporeSigma
Description: (4-MU-a-D-Glc). Crystalline solid. PROTECT FROM LIGHT. Substrate for fluorogenic assay of a-glucosidase. Has been used in studies of Pompe disease. Purity: >= 98% by enzymatic assay. Contaminants: 4-methylumbelliferone: <0.05%; b-anomer: <0.02%.