    Sort by:
5-Bromo-4-chloro-3-indoxyl-N-acetyl-beta-D-glucosaminide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   5-Bromo-4-chloro-3-indolyl N-acetyl-β-D-glucosaminide is a histochemical substrate for N-Acetylglucosaminidase. It releases an insoluble blue chromoph...