    Sort by:
  • Search Within Results
LY3023414
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   LY3023414 is a small molecule that has been shown in vitro to be a selective ATP-competitive inhibitor of PI3Kα and mTOR, DNA-PK, and other class I PI...