    Sort by:
  • Search Within Results
GDC-0349
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   GDC-0349 is a potent and selective ATP-competitive inhibitor of mTOR with Ki value of 3.8 nM.