    Sort by:
  • Search Within Results
PKI-402
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   PKI-402 is a potent dual inhibitor of PI3K/mTOR with IC50 values of 2 nM/7 nM/16 nM/14 nM and 3 nM for PI3Kα/β/γ/δ and mTOR, respectively.