    Sort by:
CP-466722
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   CP-466722 is a selective inhibitor of ATM kinase with IC50 value of 0.41 μM.