    Sort by:
  • Search Within Results
OSI-930
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   OSI-930 is a potent inhibitor of receptor tyrosine kinases with IC50 values of 80 nM, 9 nM, 8 nM, 15 nM, 41 nM and 22 nM for c-Kit, KDR, Flt, CSF-1R, ...