    Sort by:
You Searched For:
PH-797804
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   PH-797804 is a novel pyridinone inhibitor of p38α with IC50 of 26 nM; 4-fold more selective versus p38β.