    Sort by:
PD 169316
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   PD 169316 is a potent, cell-permeable and selective p38 MAP kinase inhibitor (IC50 = 89 nM).