    Sort by:
  • Search Within Results
JNK-IN-7
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   JNK-IN-7 is a selective JNK inhibitor with IC50 values of 1.54 nM, 1.99 nM, 0.75 nM to JNK1, JNK2, JNK3, respectively. It also inhibits phosphorylatio...