    Sort by:
  • Search Within Results
FR180204
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   FR180204 is a potent and selective ATP-competitive inhibitor of ERK1 and ERK2 with Ki value of 0.31 uM and 0.14 uM respectively; has IC50 values of 0....