    Sort by:
  • Search Within Results
PF-4989216
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   PF-4989216 is a novel potent and selective PI3K inhibitor. PF-4989216 inhibits PI3Kalpha (Ki=0.6 nM) and mTOR (Ki=1440 nM).