    Sort by:
You Searched For:
LMK-235
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   LMK235 is a HDAC inhibitor, previously shown to be a novel and specific inhibitor of human HDAC4 and 5, with IC50 values of 0.49 μM (A2780) and 0.32 μ...