    Sort by:
  • Search Within Results
PHA-767491(HCl)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   PHA-767491 is a potent and selective ATP-competitive dual cdc7/cdk9 inhibitor.