    Sort by:
  • Search Within Results
WAY-362450
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   WAY-362450 is a highly potent, selective, and orally active farnesoid X receptor (FXR) agonist with an EC50 of 4 nM.